gRNA Design Details
- Target DNA Sequence
- ATGTAGAGAGTGTAGCGCCG
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 142464427 - 142464449 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRB1
- Entrez Gene ID
-
575
- Gene Name
- adhesion G protein-coupled receptor B1
- Gene Aliases
-
BAI1, GDAIF
- Chromosome Location
-
Chr.8: 142449703 - 142545009 on GRCh38
- UniGene ID
- Hs.194654
- Species
-
Human