gRNA Design Details

Target DNA Sequence
GTTCATACCTGAGGATCCGA
PAM Sequence
AGG
Target locus
Chr.19: 14396442 - 14396464 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 94.7/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRE5
Entrez Gene ID
976
Gene Name
adhesion G protein-coupled receptor E5
Gene Aliases
CD97, TM7LN1
Chromosome Location
Chr.19: 14381144 - 14408725 on GRCh38
UniGene ID
Hs.466039
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001025160.2 NP_001020331.1 5