gRNA Design Details
- Target DNA Sequence
- CGTGTCTCCGACAGCTCAGA
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 48490895 - 48490917 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
76.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Rbm7
- Entrez Gene ID
-
67010
- Gene Name
- RNA binding motif protein 7
- Gene Aliases
-
1200007M24Rik, 1500011D06Rik, AU041934, AW554393
- Chromosome Location
-
Chr.9: 48488697 - 48495341 on GRCm38
- UniGene ID
- Mm.76911
- Species
-
Mouse