gRNA Design Details

Target DNA Sequence
TCGTCGATTAAAACCGCGAT
PAM Sequence
CGG
Target locus
Chr.11: 111766270 - 111766292 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 97.818/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PPP2R1B
Entrez Gene ID
5519
Gene Name
protein phosphatase 2 scaffold subunit Abeta
Gene Aliases
PP2A-Abeta, PR65B
Chromosome Location
Chr.11: 111694569 - 111766445 on GRCh38
UniGene ID
Hs.269128
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001177562.1 NP_001171033.1 1
NM_001177563.1 NP_001171034.1 1
NM_002716.4 NP_002707.3 1
NM_181699.2 NP_859050.1 1
NM_181700.1 NP_859051.1 1
XM_017017958.1 XP_016873447.1 1
XM_017017959.1 XP_016873448.1 1
XM_017017960.1 XP_016873449.1 1
XM_017017961.1 XP_016873450.1 1