gRNA Design Details
- Target DNA Sequence
- TGGTCAGGTTGATGCCACTC
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 99566334 - 99566356 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
82.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Krtap3-1
- Entrez Gene ID
-
69473
- Gene Name
- keratin associated protein 3-1
- Gene Aliases
-
2310003K17Rik, KAP3.1, KRTAP3.1
- Chromosome Location
-
Chr.11: 99566027 - 99566630 on GRCm38
- UniGene ID
- Mm.158281
- Species
-
Mouse