gRNA Design Details
- Target DNA Sequence
- CAGCCTTAGCACTTCTGCGA
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 44746221 - 44746243 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gtpbp8
- Entrez Gene ID
-
66067
- Gene Name
- GTP-binding protein 8 (putative)
- Gene Aliases
-
0610037H22Rik
- Chromosome Location
-
Chr.16: 44738879 - 44746377 on GRCm38
- UniGene ID
- Mm.34520
- Species
-
Mouse