gRNA Design Details
- Target DNA Sequence
- GAAGCCTCCGAATATGAGCG
- PAM Sequence
- CGG
- Target locus
-
Chr.5: 53267269 - 53267291 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.14/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Smim20
- Entrez Gene ID
-
66278
- Gene Name
- small integral membrane protein 20
- Gene Aliases
-
1110067B18Rik, 1810013D10Rik, N28078
- Chromosome Location
-
Chr.5: 53267106 - 53278540 on GRCm38
- UniGene ID
- Mm.27900
- Species
-
Mouse