gRNA Design Details
- Target DNA Sequence
- GGAGCGGTAAGATTGGTTTC
- PAM Sequence
- GGG
- Application
- Gene Knock-Out
Score:
92.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Nkap
- Entrez Gene ID
-
67050
- Gene Name
- NFKB activating protein
- Gene Aliases
-
2610020O08Rik, AA987160, AI849147, AL024236
- Chromosome Location
-
Chr.X: 37126763 - 37150746 on GRCm38
- UniGene ID
- Mm.55422
- Species
-
Mouse