gRNA Design Details
- Target DNA Sequence
- TACAAACGCCTGGACTGACT
- PAM Sequence
- CGG
- Target locus
-
Chr.9: 63616833 - 63616855 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Aagab
- Entrez Gene ID
-
66939
- Gene Name
- alpha- and gamma-adaptin binding protein
- Gene Aliases
-
2310007F21Rik, p34
- Chromosome Location
-
Chr.9: 63602629 - 63641897 on GRCm38
- UniGene ID
- Mm.34338
- Species
-
Mouse