gRNA Design Details
- Target DNA Sequence
- TCGATTACAGCTGTGCTTGT
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 155959776 - 155959798 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Cep350
- Entrez Gene ID
-
74081
- Gene Name
- centrosomal protein 350
- Gene Aliases
-
4933409L06Rik, 6430546F08Rik
- Chromosome Location
-
Chr.1: 155844964 - 155974435 on GRCm38
- UniGene ID
- Mm.260247
- Species
-
Mouse