gRNA Design Details

Target DNA Sequence
TGACGCAAACTATGATGGAA
PAM Sequence
AGG
Target locus
Chr.2: 27024173 - 27024195 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 68.02/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MAPRE3
Entrez Gene ID
22924
Gene Name
microtubule associated protein RP/EB family member 3
Gene Aliases
EB3, EBF3, EBF3-S, RP3
Chromosome Location
Chr.2: 26970371 - 27027219 on GRCh38
UniGene ID
Hs.515860
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001303050.1 NP_001289979.1 4
NM_012326.3 NP_036458.2 4
XM_006711967.3 XP_006712030.1 4
XM_011532700.1 XP_011531002.1 4
XM_017003597.1 XP_016859086.1 4