gRNA Design Details
- Target DNA Sequence
- TTCCTCAAGTGAACACAGCC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 30862533 - 30862555 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
81.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mapre3
- Entrez Gene ID
-
100732
- Gene Name
- microtubule-associated protein, RP/EB family, member 3
- Gene Aliases
-
AI790651, EB2, EB3
- Chromosome Location
-
Chr.5: 30814614 - 30866106 on GRCm38
- UniGene ID
- Mm.332295
- Species
-
Mouse