gRNA Design Details
- Target DNA Sequence
- GCAGCCCTTGGGATAATTAT
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 86671821 - 86671843 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.44/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmprss11b
- Entrez Gene ID
-
319875
- Gene Name
- transmembrane protease, serine 11B
- Gene Aliases
-
9930019B18Rik, Tmprss11bnl
- Chromosome Location
-
Chr.5: 86659522 - 86676298 on GRCm38
- UniGene ID
- Mm.253994
- Species
-
Mouse