gRNA Design Details
- Target DNA Sequence
- TACCAAGACCTGCAGCCCGT
- PAM Sequence
- TGG
- Target locus
-
Chr.15: 89140372 - 89140394 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mapk12
- Entrez Gene ID
-
29857
- Gene Name
- mitogen-activated protein kinase 12
- Gene Aliases
-
AW123708, Erk6, P38gamma, Prkm12, Sapk3
- Chromosome Location
-
Chr.15: 89130584 - 89141593 on GRCm38
- UniGene ID
- Mm.38343
- Species
-
Mouse