gRNA Design Details
- Target DNA Sequence
- CGTGGCGCATGTGTTTGAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 89140133 - 89140155 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.24/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mapk12
- Entrez Gene ID
-
29857
- Gene Name
- mitogen-activated protein kinase 12
- Gene Aliases
-
AW123708, Erk6, P38gamma, Prkm12, Sapk3
- Chromosome Location
-
Chr.15: 89130584 - 89141593 on GRCm38
- UniGene ID
- Mm.38343
- Species
-
Mouse