gRNA Design Details
- Target DNA Sequence
- CCAAGCTAATGAGCGTGTGA
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 120660791 - 120660813 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.5/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Emc8
- Entrez Gene ID
-
18117
- Gene Name
- ER membrane protein complex subunit 8
- Gene Aliases
-
Cox4nb, Fam158b, Noc4
- Chromosome Location
-
Chr.8: 120653914 - 120668112 on GRCm38
- UniGene ID
- Mm.2136
- Species
-
Mouse