gRNA Design Details
- Target DNA Sequence
- ATGGAATAGCGCTGTCCCTC
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 106301174 - 106301196 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
82.71/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gh
- Entrez Gene ID
-
14599
- Gene Name
- growth hormone
- Gene Aliases
-
Gh1
- Chromosome Location
-
Chr.11: 106300261 - 106303704 on GRCm38
- UniGene ID
- Mm.343934
- Species
-
Mouse