gRNA Design Details
- Target DNA Sequence
- TGGTCAAGATCTTCACAGAA
- PAM Sequence
- GGG
- Target locus
-
Chr.19: 32799914 - 32799936 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
81.86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pten
- Entrez Gene ID
-
19211
- Gene Name
- phosphatase and tensin homolog
- Gene Aliases
-
2310035O07Rik, A130070J02Rik, AI463227, B430203M17Rik, MMAC1, TEP1
- Chromosome Location
-
Chr.19: 32757577 - 32826160 on GRCm38
- UniGene ID
- Mm.245395
- Species
-
Mouse