gRNA Design Details
- Target DNA Sequence
- AAAAGGAGATATCAAGAGGA
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 87864506 - 87864528 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
56.25/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PTEN
- Entrez Gene ID
-
5728
- Gene Name
- phosphatase and tensin homolog
- Gene Aliases
-
10q23del, BZS, CWS1, DEC, GLM2, MHAM, MMAC1, PTEN1, TEP1
- Chromosome Location
-
Chr.10: 87863438 - 87971930 on GRCh38
- UniGene ID
- Hs.500466
- Species
-
Human