gRNA Design Details
- Target DNA Sequence
- AGAAGCGGAAGTCTTCTCGG
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 95003267 - 95003289 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.25/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrg1
- Entrez Gene ID
-
14766
- Gene Name
- adhesion G protein-coupled receptor G1
- Gene Aliases
-
Cyt28, Gpr56, TM7LN4, TM7XN1
- Chromosome Location
-
Chr.8: 94974756 - 95014208 on GRCm38
- UniGene ID
- Mm.290834
- Species
-
Mouse