gRNA Design Details
- Target DNA Sequence
- AGCGGTGCTTAGAGCAACTG
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 55328448 - 55328470 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.8/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mxd3
- Entrez Gene ID
-
17121
- Gene Name
- Max dimerization protein 3
- Gene Aliases
-
4631412E13Rik, Mad3, bHLHc13
- Chromosome Location
-
Chr.13: 55325168 - 55329770 on GRCm38
- UniGene ID
- Mm.20350
- Species
-
Mouse