gRNA Design Details

Target DNA Sequence
TCAAACCTGTGGTGTAGGAC
PAM Sequence
AGG
Target locus
Chr.8: 55762923 - 55762945 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 83.43/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TMEM68
Entrez Gene ID
137695
Gene Name
transmembrane protein 68
Gene Aliases
-
Chromosome Location
Chr.8: 55738744 - 55773407 on GRCh38
UniGene ID
Hs.420076
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001286657.1 NP_001273586.1 2
NM_001286660.1 NP_001273589.1 3
NM_152417.2 NP_689630.1 3
XM_005251150.1 XP_005251207.1 3
XM_011517454.1 XP_011515756.1 3
XM_011517455.2 XP_011515757.1 3