gRNA Design Details

Target DNA Sequence
GCGGATCGCACTCTCTTTGT
PAM Sequence
GGG
Target locus
Chr.11: 114400693 - 114400715 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.56/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
RBM7
Entrez Gene ID
10179
Gene Name
RNA binding motif protein 7
Gene Aliases
-
Chromosome Location
Chr.11: 114400529 - 114410610 on GRCh38
UniGene ID
Hs.7527
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001286045.1 NP_001272974.1 1
NM_001286046.1 NP_001272975.1 1
NM_001286047.1 NP_001272976.1 1
NM_001286048.1 NP_001272977.1 1
NM_016090.3 NP_057174.1 1