gRNA Design Details
- Target DNA Sequence
- GCGTATCTTTGACCCAAGCC
- PAM Sequence
- CGG
- Target locus
-
Chr.3: 112991203 - 112991225 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.02/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GTPBP8
- Entrez Gene ID
-
29083
- Gene Name
- GTP binding protein 8 (putative)
- Gene Aliases
-
HSPC135
- Chromosome Location
-
Chr.3: 112989275 - 113001966 on GRCh38
- UniGene ID
- Hs.127496
- Species
-
Human