gRNA Design Details
- Target DNA Sequence
- CAGTGGCTTTTACCGCCAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.22: 50261464 - 50261486 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.53/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MAPK12
- Entrez Gene ID
-
6300
- Gene Name
- mitogen-activated protein kinase 12
- Gene Aliases
-
ERK-6, ERK3, ERK6, MAPK 12, P38GAMMA, PRKM12, SAPK-3, SAPK3
- Chromosome Location
-
Chr.22: 50252902 - 50261759 on GRCh38
- UniGene ID
- Hs.432642
- Species
-
Human