gRNA Design Details
- Target DNA Sequence
- ATGCTAACTCCTTCTGCTGG
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 68234552 - 68234574 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMPRSS11B
- Entrez Gene ID
-
132724
- Gene Name
- transmembrane protease, serine 11B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 68226653 - 68245694 on GRCh38
- UniGene ID
- Hs.407122
- Species
-
Human