gRNA Design Details
- Target DNA Sequence
- TCAAGCTAATGAGCGAGTAA
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 85788981 - 85789003 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.31/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EMC8
- Entrez Gene ID
-
10328
- Gene Name
- ER membrane protein complex subunit 8
- Gene Aliases
-
C16orf2, C16orf4, COX4NB, FAM158B, NOC4
- Chromosome Location
-
Chr.16: 85778624 - 85799744 on GRCh38
- UniGene ID
- Hs.173162
- Species
-
Human