gRNA Design Details
- Target DNA Sequence
- ACTGTCAGAATCCCTGGCCG
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 120220753 - 120220775 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.57/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmbim4
- Entrez Gene ID
-
68212
- Gene Name
- transmembrane BAX inhibitor motif containing 4
- Gene Aliases
-
0610007H07Rik, AU022431
- Chromosome Location
-
Chr.10: 120208826 - 120224897 on GRCm38
- UniGene ID
- Mm.157426
- Species
-
Mouse