gRNA Design Details

Target DNA Sequence
TGCTTCAGACCTGTTTCCCG
PAM Sequence
CGG
Target locus
Chr.15: 82689190 - 82689212 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 88.21/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AP3B2
Entrez Gene ID
8120
Gene Name
adaptor related protein complex 3 beta 2 subunit
Gene Aliases
NAPTB
Chromosome Location
Chr.15: 82659281 - 82710112 on GRCh38
UniGene ID
Hs.199593
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001278511.1 NP_001265440.1 3
NM_001278512.1 NP_001265441.1 3
NM_004644.4 NP_004635.2 3
XM_017022640.1 XP_016878129.1 3
XM_017022641.1 XP_016878130.1 3