gRNA Design Details

Target DNA Sequence
GGATGCACTTTCTCAAACCA
PAM Sequence
AGG
Target locus
Chr.1: 179987259 - 179987281 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 86.57/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CEP350
Entrez Gene ID
9857
Gene Name
centrosomal protein 350
Gene Aliases
CAP350, GM133
Chromosome Location
Chr.1: 179954773 - 180114880 on GRCh38
UniGene ID
Hs.413045
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_014810.4 NP_055625.4 3
XM_011510191.2 XP_011508493.1 3
XM_011510198.2 XP_011508500.1 3
XM_017002929.1 XP_016858418.1 3
XM_017002930.1 XP_016858419.1 3
XM_017002931.1 XP_016858420.1 3