gRNA Design Details
- Target DNA Sequence
- TGCCCAGGTTGATAGCATTG
- PAM Sequence
- TGG
- Target locus
-
Chr.15: 67208562 - 67208584 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AAGAB
- Entrez Gene ID
-
79719
- Gene Name
- alpha- and gamma-adaptin binding protein
- Gene Aliases
-
KPPP1, PPKP1, PPKP1A, p34
- Chromosome Location
-
Chr.15: 67200675 - 67255198 on GRCh38
- UniGene ID
- Hs.254642
- Species
-
Human