gRNA Design Details
- Target DNA Sequence
- AGAGAAGCTTCGGCATGCCG
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 125192407 - 125192429 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.8/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mrpl51
- Entrez Gene ID
-
66493
- Gene Name
- mitochondrial ribosomal protein L51
- Gene Aliases
-
2610511M02Rik, CDA09, HSPC241, Mrp64
- Chromosome Location
-
Chr.6: 125192200 - 125194392 on GRCm38
- UniGene ID
- Mm.138749
- Species
-
Mouse