gRNA Design Details
- Target DNA Sequence
- TTTGATAAAAGCTGCCGCTG
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 41008989 - 41009011 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- KRTAP3-1
- Entrez Gene ID
-
83896
- Gene Name
- keratin associated protein 3-1
- Gene Aliases
-
KAP3.1, KRTAP3.1
- Chromosome Location
-
Chr.17: 41008521 - 41009114 on GRCh38
- UniGene ID
- Hs.307027
- Species
-
Human