gRNA Design Details
- Target DNA Sequence
- GCAGCAACGCTAGAAGACAG
- PAM Sequence
- CGG
- Target locus
-
Chr.20: 6770141 - 6770163 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.8/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BMP2
- Entrez Gene ID
-
650
- Gene Name
- bone morphogenetic protein 2
- Gene Aliases
-
BDA2, BMP2A
- Chromosome Location
-
Chr.20: 6767700 - 6780280 on GRCh38
- UniGene ID
- Hs.73853
- Species
-
Human