gRNA Design Details
- Target DNA Sequence
- ACCCTTGGCCAGCAACATCC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 177311803 - 177311825 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MXD3
- Entrez Gene ID
-
83463
- Gene Name
- MAX dimerization protein 3
- Gene Aliases
-
BHLHC13, MAD3, MYX
- Chromosome Location
-
Chr.5: 177305500 - 177312291 on GRCh38
- UniGene ID
- Hs.743332
- Species
-
Human