gRNA Design Details
- Target DNA Sequence
- GTTGAAGTCGTCCTCGATCG
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 66169898 - 66169920 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
96.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMBIM4
- Entrez Gene ID
-
51643
- Gene Name
- transmembrane BAX inhibitor motif containing 4
- Gene Aliases
-
CGI-119, GAAP, LFG4, S1R, ZPRO
- Chromosome Location
-
Chr.12: 66136936 - 66170072 on GRCh38
- UniGene ID
- Hs.505934
- Species
-
Human