gRNA Design Details
- Target DNA Sequence
- CTTCCGGCCCCTAATGAGAT
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 25914391 - 25914413 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.56/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SMIM20
- Entrez Gene ID
-
389203
- Gene Name
- small integral membrane protein 20
- Gene Aliases
-
C4orf52
- Chromosome Location
-
Chr.4: 25914192 - 25929879 on GRCh38
- UniGene ID
- Hs.479386
- Species
-
Human