gRNA Design Details
- Target DNA Sequence
- GCCGTAATAGACTGACGAGG
- PAM Sequence
- AGG
- Application
- Gene Knockout
Score:
97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NKAP
- Entrez Gene ID
-
79576
- Gene Name
- NFKB activating protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 119925050 - 119943772 on GRCh38
- UniGene ID
- Hs.522771
- Species
-
Human