gRNA Design Details

Target DNA Sequence
TGATCGTTGGAACGAGAAAA
PAM Sequence
GGG
Target locus
Chr.12: 6492898 - 6492920 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 90.46/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MRPL51
Entrez Gene ID
51258
Gene Name
mitochondrial ribosomal protein L51
Gene Aliases
CDA09, HSPC241, MRP64, bMRP64
Chromosome Location
Chr.12: 6492150 - 6493305 on GRCh38
UniGene ID
Hs.55847
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_016497.3 NP_057581.2 2