gRNA Design Details
- Target DNA Sequence
- TTCGGAGTGTATGACAACAT
- PAM Sequence
- CGG
- Target locus
-
Chr.12: 6492870 - 6492892 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.08/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MRPL51
- Entrez Gene ID
-
51258
- Gene Name
- mitochondrial ribosomal protein L51
- Gene Aliases
-
CDA09, HSPC241, MRP64, bMRP64
- Chromosome Location
-
Chr.12: 6492150 - 6493305 on GRCh38
- UniGene ID
- Hs.55847
- Species
-
Human