gRNA Design Details
- Target DNA Sequence
- CTTCTCCTCACTAAGGCCAT
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 19132727 - 19132749 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
84.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMEM161A
- Entrez Gene ID
-
54929
- Gene Name
- transmembrane protein 161A
- Gene Aliases
-
AROS-29
- Chromosome Location
-
Chr.19: 19119616 - 19138501 on GRCh38
- UniGene ID
- Hs.631629
- Species
-
Human