Gene Transcripts

Gene Symbol
MAP1A
Entrez Gene ID
4130
Gene Name
microtubule associated protein 1A
Gene Aliases
MAP1L, MTAP1A
Chromosome Location
Chr. 15: 43510946 - 43531620 on Build GRCh38
UniGene ID
Hs.194301
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002373.5 NP_002364.5 4 1000
XM_017022189.1 XP_016877678.1 5 1290
GenBank mRNA AF200415.2 - 2 732
AK291052.1 - 4 972
BC055312.1 - 4 969
BC094702.1 - 4 959
U38292.1 - 1 533
U80458.1 - 1 302

View Full Product Details