Gene Transcripts

Gene Symbol
HMBOX1
Entrez Gene ID
79618
Gene Name
homeobox containing 1
Gene Aliases
HNF1LA, HOT1, PBHNF
Chromosome Location
Chr. 8: 28890370 - 29064939 on Build GRCh38
UniGene ID
Hs.563560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135726.2 NP_001129198.1 6 1100
NM_001324382.1 NP_001311311.1 6 960
NM_001324383.1 NP_001311312.1 6 1100
NM_001324384.1 NP_001311313.1 6 1103
NM_001324385.1 NP_001311314.1 6 1100
NM_001324386.1 NP_001311315.1 6 1100
NM_001324387.1 NP_001311316.1 6 1100
NM_001324388.1 NP_001311317.1 6 960
NM_001324389.1 NP_001311318.1 6 1100
NM_001324390.1 NP_001311319.1 6 1103
NM_001324391.1 NP_001311320.1 6 1100
NM_001324392.1 NP_001311321.1 6 1103
NM_001324393.1 NP_001311322.1 7 1216
NM_001324394.1 NP_001311323.1 6 1100
NM_001324395.1 NP_001311324.1 6 1100
NM_024567.4 NP_078843.2 7 1134
NR_136757.1 5 1014
NR_136758.1 6 1100
XM_005273635.1 XP_005273692.1 6 1076
XM_005273641.4 XP_005273698.1 6 1076
XM_006716398.2 XP_006716461.1 6 1076
XM_017013823.1 XP_016869312.1 6 1108
XM_017013824.1 XP_016869313.1 6 898
XM_017013825.1 XP_016869314.1 6 1076
GenBank mRNA AK025269.1 - 4 565
AK290683.1 - 6 1076
AK295320.1 - 5 739
AK310959.1 - 6 1066
AY522342.1 - 6 845
BC009259.2 - 4 518
BC069242.1 - 7 1134 5928365
DQ153248.1 - 4 446
DQ269478.1 - 6 845

View Full Product Details