Gene Transcripts
- Gene Symbol
- THOC7
- Entrez Gene ID
- 80145
- Gene Name
- THO complex 7
- Gene Aliases
- NIF3L1BP1, fSAP24, hTREX30
- Chromosome Location
- Chr. 3: 63833870 - 63864478 on Build GRCh38
- UniGene ID
- Hs.288151
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285387.2 |
NP_001272316.1
|
5, 6 |
502 |
|
NM_001285404.1 |
NP_001272333.1
|
6, 7 |
751 |
|
NM_025075.3 |
NP_079351.2
|
6, 7 |
620 |
|
XM_006713339.3 |
XP_006713402.1
|
6, 7 |
569 |
|
GenBank mRNA |
AF104914.1 |
- |
5, 6 |
468 |
|
AK027098.1 |
- |
6, 7 |
530 |
|
AK225110.1 |
- |
6, 7 |
508 |
|
BC020599.2 |
- |
5, 6 |
468 |
|
BC065012.1 |
- |
6, 7 |
490 |
6167514 |
CR457373.1 |
- |
6, 7 |
487 |
|
View Full Product Details