Gene Transcripts

Gene Symbol
TSPYL5
Entrez Gene ID
85453
Gene Name
TSPY like 5
Gene Aliases
Chromosome Location
Chr. 8: 97273486 - 97277948 on Build GRCh38
UniGene ID
Hs.173094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033512.2 NP_277047.2 1 1038
GenBank mRNA AB051537.1 - 1 977
AK091445.1 - 1 1054
BC045630.1 - 1 1038 5273220

View Full Product Details