Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143830.1 | NP_001137302.1 | 2 | 356 | |
NM_005256.3 | NP_005247.1 | 1 | 147 | ||
NM_177553.2 | NP_808221.1 | 2 | 298 | ||
XM_011519971.2 | XP_011518273.1 | 2 | 356 | ||
XM_011519972.2 | XP_011518274.1 | 2 | 2351 | ||
XM_011519975.1 | XP_011518277.1 | 2 | 356 | ||
XM_011519978.2 | XP_011518280.1 | 2 | 356 | ||
XM_017017528.1 | XP_016873017.1 | 2 | 1048 | ||
XM_017017529.1 | XP_016873018.1 | 2 | 193 | ||
XM_017017530.1 | XP_016873019.1 | 2 | 386 | ||
XM_017017531.1 | XP_016873020.1 | 2 | 207 | ||
XM_017017532.1 | XP_016873021.1 | 2 | 356 | ||
GenBank mRNA | AK313734.1 | - | 2 | 362 | |
AU099094.1 | - | 1 | 147 | ||
BC013326.1 | - | 2 | 302 | ||
BC040470.1 | - | 2 | 298 | 4280621 | |
CR450285.1 | - | 1 | 50 | ||
DA345287.1 | - | 2 | 356 | ||
U95032.1 | - | 1 | 116 |