Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000173.6 | NP_000164.5 | 2 | 1993 | |
GenBank mRNA | BC027955.1 | - | 3 | 1893 | |
J02940.1 | - | 3 | 1860 | ||
KC120774.1 | - | 2 | 1970 | ||
KC120775.1 | - | 2 | 1969 | ||
KC120776.1 | - | 2 | 1970 |