Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 117311037-117311583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROS1
Entrez Gene ID
6098
Gene Name
ROS proto-oncogene 1, receptor tyrosine kinase
Gene Aliases
MCF3, ROS, c-ros-1
UniGene
Hs.1041
Gene Chromosome Location
Chr.6: 117287918-117426065 on Build GRCh38
Transcripts
NM_002944.2 XM_006715548.3 XM_011536049.2 XM_011536050.2 XM_011536051.2 XM_011536052.2 XM_011536053.2 XM_011536054.2 XM_017011172.1 XM_017011173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6950684 c.6159G>T p.L2053F Chr.6: 117311094-117311094 on Build GRCh38
1072517 c.6158T>C p.L2053S Chr.6: 117311095-117311095 on Build GRCh38
6172042 c.6196G>T p.G2066C Chr.6: 117311057-117311057 on Build GRCh38
3857780 c.6125G>A p.G2042D Chr.6: 117311110-117311110 on Build GRCh38
6975258 c.6211G>A p.E2071K Chr.6: 117311042-117311042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575072554 Chr.6: 117311316-117311316 on Build GRCh38
rs1037599478 Chr.6: 117311395-117311395 on Build GRCh38
rs949081219 Chr.6: 117311271-117311271 on Build GRCh38
rs1030742170 Chr.6: 117311491-117311491 on Build GRCh38
rs959844455 Chr.6: 117311291-117311291 on Build GRCh38
rs932901940 Chr.6: 117311334-117311334 on Build GRCh38
rs979977164 Chr.6: 117311254-117311254 on Build GRCh38
rs149651586 Chr.6: 117311326-117311326 on Build GRCh38
rs776308471 Chr.6: 117311076-117311076 on Build GRCh38
rs557368820 Chr.6: 117311328-117311328 on Build GRCh38
rs891302592 Chr.6: 117311353-117311353 on Build GRCh38
rs182672503 Chr.6: 117311180-117311180 on Build GRCh38
rs193121244 Chr.6: 117311065-117311065 on Build GRCh38
rs1001426865 Chr.6: 117311478-117311478 on Build GRCh38
rs1030253447 Chr.6: 117311179-117311179 on Build GRCh38
rs573019683 Chr.6: 117311533-117311533 on Build GRCh38
rs529164211 Chr.6: 117311098-117311098 on Build GRCh38
rs751532443 Chr.6: 117311097-117311097 on Build GRCh38
rs957864391 Chr.6: 117311556-117311556 on Build GRCh38
rs191629132 Chr.6: 117311304-117311304 on Build GRCh38
rs201894511 Chr.6: 117311093-117311093 on Build GRCh38
rs776060632 Chr.6: 117311060-117311060 on Build GRCh38
rs372627493 Chr.6: 117311542-117311542 on Build GRCh38
rs986028893 Chr.6: 117311546-117311546 on Build GRCh38
rs558109082 Chr.6: 117311303-117311303 on Build GRCh38
rs575638193 Chr.6: 117311398-117311398 on Build GRCh38
rs768701106 Chr.6: 117311203-117311203 on Build GRCh38
rs1015639649 Chr.6: 117311178-117311178 on Build GRCh38
rs760630574 Chr.6: 117311519-117311519 on Build GRCh38
rs368003464 Chr.6: 117311073-117311073 on Build GRCh38
rs9489121 Chr.6: 117311443-117311443 on Build GRCh38
rs764825728 Chr.6: 117311080-117311080 on Build GRCh38
rs921584442 Chr.6: 117311327-117311327 on Build GRCh38
rs1008361386 Chr.6: 117311393-117311393 on Build GRCh38
rs767378516 Chr.6: 117311146-117311146 on Build GRCh38
rs1043104807 Chr.6: 117311061-117311061 on Build GRCh38
rs144370245 Chr.6: 117311541-117311541 on Build GRCh38
rs766298478 Chr.6: 117311094-117311094 on Build GRCh38
rs1032230610 Chr.6: 117311547-117311547 on Build GRCh38
rs969974037 Chr.6: 117311223-117311223 on Build GRCh38
rs997568298 Chr.6: 117311455-117311455 on Build GRCh38
rs184313070 Chr.6: 117311554-117311554 on Build GRCh38
rs947120863 Chr.6: 117311315-117311315 on Build GRCh38
rs150410491 Chr.6: 117311149-117311149 on Build GRCh38
rs1014938952 Chr.6: 117311174-117311174 on Build GRCh38
rs1035535148 Chr.6: 117311501-117311501 on Build GRCh38
rs779050446 Chr.6: 117311154-117311154 on Build GRCh38
rs761415842 Chr.6: 117311077-117311077 on Build GRCh38
rs758489691 Chr.6: 117311169-117311169 on Build GRCh38
rs576755852 Chr.6: 117311151-117311151 on Build GRCh38
rs73564794 Chr.6: 117311288-117311288 on Build GRCh38
rs897865555 Chr.6: 117311447-117311447 on Build GRCh38
rs925096542 Chr.6: 117311206-117311206 on Build GRCh38
rs962032772 Chr.6: 117311177-117311177 on Build GRCh38
rs986017936 Chr.6: 117311204-117311204 on Build GRCh38
rs1027016779 Chr.6: 117311433-117311433 on Build GRCh38
rs186827380 Chr.6: 117311245-117311245 on Build GRCh38
rs111617088 Chr.6: 117311397-117311397 on Build GRCh38
rs913618089 Chr.6: 117311270-117311270 on Build GRCh38
rs1034471676 Chr.6: 117311083-117311083 on Build GRCh38
rs771784942 Chr.6: 117311330-117311330 on Build GRCh38
rs564090141 Chr.6: 117311530-117311530 on Build GRCh38
rs772539932 Chr.6: 117311081-117311081 on Build GRCh38
rs1049980735 Chr.6: 117311347-117311347 on Build GRCh38
rs1020516478 Chr.6: 117311532-117311532 on Build GRCh38
rs113375607 Chr.6: 117311202-117311202 on Build GRCh38
rs746002195 Chr.6: 117311162-117311162 on Build GRCh38
rs987899940 Chr.6: 117311267-117311267 on Build GRCh38

View Full Product Details