Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93068831-93069333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts
XM_017021263.1 XM_017021264.1 NR_002808.2

Gene Symbol
ITPK1-AS1
Entrez Gene ID
319085
Gene Name
ITPK1 antisense RNA 1
Gene Aliases
C14orf85, ITPK1-AS, ITPK1AS, NCRNA00203
UniGene
Hs.612888
Gene Chromosome Location
Chr.14: 93067452-93072152 on Build GRCh38
Transcripts
XM_017021263.1 XM_017021264.1 NR_002808.2

SNP Detail

SNP ID Chromosome Location
rs769008996 Chr.14: 93068865-93068865 on Build GRCh38
rs556436682 Chr.14: 93068877-93068877 on Build GRCh38
rs935042788 Chr.14: 93069284-93069284 on Build GRCh38
rs3814827 Chr.14: 93068852-93068852 on Build GRCh38
rs998661567 Chr.14: 93068852-93068853 on Build GRCh38
rs989251007 Chr.14: 93069287-93069287 on Build GRCh38
rs909060516 Chr.14: 93068908-93068908 on Build GRCh38
rs150099213 Chr.14: 93069071-93069071 on Build GRCh38
rs775993142 Chr.14: 93068851-93068851 on Build GRCh38
rs375387974 Chr.14: 93069224-93069224 on Build GRCh38
rs970798097 Chr.14: 93069231-93069231 on Build GRCh38
rs147596850 Chr.14: 93068967-93068967 on Build GRCh38
rs766256082 Chr.14: 93069299-93069299 on Build GRCh38
rs1023587441 Chr.14: 93069226-93069226 on Build GRCh38
rs773957146 Chr.14: 93068892-93068892 on Build GRCh38
rs963239036 Chr.14: 93068924-93068924 on Build GRCh38
rs767264600 Chr.14: 93069058-93069058 on Build GRCh38
rs1021497249 Chr.14: 93068928-93068928 on Build GRCh38
rs369048168 Chr.14: 93068984-93068985 on Build GRCh38
rs1014970926 Chr.14: 93069018-93069018 on Build GRCh38
rs531812081 Chr.14: 93069194-93069194 on Build GRCh38
rs1001357048 Chr.14: 93069245-93069245 on Build GRCh38
rs923613533 Chr.14: 93069272-93069272 on Build GRCh38
rs1035970763 Chr.14: 93069258-93069258 on Build GRCh38
rs901159284 Chr.14: 93069082-93069082 on Build GRCh38
rs983210646 Chr.14: 93068907-93068907 on Build GRCh38
rs141959392 Chr.14: 93069004-93069004 on Build GRCh38
rs1053215236 Chr.14: 93069150-93069150 on Build GRCh38
rs1025603724 Chr.14: 93069118-93069118 on Build GRCh38
rs990772992 Chr.14: 93068949-93068949 on Build GRCh38
rs1005607180 Chr.14: 93069190-93069190 on Build GRCh38
rs753936288 Chr.14: 93069081-93069081 on Build GRCh38
rs960082838 Chr.14: 93068988-93068988 on Build GRCh38
rs527344101 Chr.14: 93069003-93069003 on Build GRCh38
rs115592701 Chr.14: 93069077-93069077 on Build GRCh38
rs944155672 Chr.14: 93069095-93069095 on Build GRCh38
rs949110622 Chr.14: 93068987-93068987 on Build GRCh38
rs981738054 Chr.14: 93069248-93069248 on Build GRCh38
rs374256018 Chr.14: 93069298-93069298 on Build GRCh38
rs891702047 Chr.14: 93068903-93068903 on Build GRCh38
rs905678965 Chr.14: 93068946-93068946 on Build GRCh38
rs370297882 Chr.14: 93069132-93069132 on Build GRCh38
rs905816834 Chr.14: 93069240-93069240 on Build GRCh38
rs571660270 Chr.14: 93069308-93069308 on Build GRCh38
rs761212713 Chr.14: 93069023-93069023 on Build GRCh38
rs188428006 Chr.14: 93068992-93068992 on Build GRCh38
rs1035431131 Chr.14: 93068968-93068968 on Build GRCh38
rs947406877 Chr.14: 93069196-93069196 on Build GRCh38
rs562771435 Chr.14: 93069083-93069083 on Build GRCh38
rs540050381 Chr.14: 93068878-93068878 on Build GRCh38
rs560892131 Chr.14: 93069020-93069020 on Build GRCh38
rs963266646 Chr.14: 93069210-93069210 on Build GRCh38
rs576288326 Chr.14: 93068904-93068904 on Build GRCh38
rs963043467 Chr.14: 93069057-93069057 on Build GRCh38

View Full Product Details