Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93070301-93070807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts
XM_017021263.1 XM_017021264.1 NR_002808.2

Gene Symbol
ITPK1-AS1
Entrez Gene ID
319085
Gene Name
ITPK1 antisense RNA 1
Gene Aliases
C14orf85, ITPK1-AS, ITPK1AS, NCRNA00203
UniGene
Hs.612888
Gene Chromosome Location
Chr.14: 93067452-93072152 on Build GRCh38
Transcripts
XM_017021263.1 XM_017021264.1 NR_002808.2

View Full Product Details